Easy grip manual crimpers
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
23398 23396 23397 23399
REST23398EA 2400 DKK
REST23398 REST23396 REST23397 REST23399
Easy grip manual crimpers
Tænger til Vials
Easy grip crimpers feature tough, light, plastic bodies with curved handles that gives hand comfort during use.
  • Bottom-pull handle allows for a steadier hold than top-push handles
  • An easily viewed adjustment knob is located at the head of the product
  • The clearly shown + and – symbols, along with directional arrows, simplify adjusting the intensity of the crimp as needed
  • Knob is designed to indicate when the crimp setting has been reached, when it fully contacts the body of the product