Dual canister sampling manifolds, stainless steel/Siltek® treated
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
24998 24999
REST24998EA 2460 DKK
REST24998 REST24999
Dual canister sampling manifolds, stainless steel/Siltek® treated
Manifolder
Dual canister sampling manifold minimises sampling variability through a single sample inlet and flow controller by which the sample is evenly collected between two canisters. Use of a single flow controller eliminates flow rate variability, as well as environmental variables common with collocated samples. It may also be used with two flow controllers to monitor individual canister vacuum.
  • Precise dimensions (W×H: 241,3×88,9 mm) provide accurate splitting of sample between two canisters
  • Minimises leaks
  • Thick-walled stainless steel tubing is rugged enough