FetalClone® I, amerikansk oprindelse, HyClone
Leverandør: HyClone products (Cytiva)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SH30080.02 SH30080.03 SH30080.02HI SH30080.03HI SH30080.03IR25
SH30080.02EA 699 DKK
SH30080.02 HYCLSH30080.03 HYCLSH30080.02HI HYCLSH30080.03HI SH30080.03IR25
FetalClone® I, amerikansk oprindelse, HyClone
Serum
FetalClone® I er optimeret til dyrkning af hybridomer, og IgG-niveauet kan sammenlignes med det, der findes i FBS.
  • IgG levels comparable with those found in FBS
  • Demonstrated cell growth performance equivalency to FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
Sterile filtered using three sequential 100 nm (0.1 µm) pore-size rated filters. Each lot undergoes extensive quality performance and analytical testing.