Vacuum gauges
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
24269 24270
REST24269EA 1380 DKK
REST24269 REST24270
Vacuum gauges
Pumper Vakuummålere
These vacuum gauges are made of stainless steel wetted surfaces with rear mount.
Suitable for passive air sampling kits.