Inlet liners, Sky®
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
23326 23305.1 23309.1 23434.5 23305.5 23312.1 23438.5 23336.5 23449.5 23312
REST23326EA 2380 DKK
REST23326 REST23305.1 REST23309.1 REST23434.5 REST23305.5 REST23312.1 REST23438.5 REST23336.5 REST23449.5 REST23312
Inlet liners, Sky®
Kromatografi forsyninger Kromatografi indløbsforinger
Sky® inlet liners with Sky® technology deactivation improve trace-level analysis that are suitable for Agilent GCs, Perkinelmer GCs, Shimadzu GCs and Thermo scientific GCs equipped with split/splitless inlets.
  • Increase accuracy and precision by preventing loss of sensitive analytes-even when using wool
  • Lower detection limits for a wide range of active compounds is achieved
  • Liner-to-liner reproducibility through consistent manufacturing and extensive testing