Ferrules for Thermo Scientific GCs
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20280 20282 20284 20283 20285
REST20280EA 321 DKK
REST20280 REST20282 REST20284 REST20283 REST20285
Ferrules for Thermo Scientific GCs
Fittings og stik
Graphite Ferrules for Vu2 union connectors.
  • High-purity, high-density graphite
  • Smoother surface and cleaner edges than conventional graphite ferrules
  • Contain no binders that can off -gas or adsorb analytes
  • Stable to 450 °C