High purity airgas VOC regulators
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
26371 26372
REST26371EA 15750 DKK
REST26371 REST26372
High purity airgas VOC regulators
Gasregulatorer
These single stage stainless steel VOC regulators features stainless steel diaphragm, two pressure gauges, CGA-180 fitting and Kel-F® seat.
  • Accurate pressure control even at low flow rates
  • Individually tested for leaks and impurities