Toggle gas valves, Parker Balston®
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22188 22189 22190 22191
REST22188EA 0 DKK
REST22188 REST22189 REST22190 REST22191
Toggle gas valves, Parker Balston®
Ventiler Luftreguleringsventiler
These shut-off toggle gas valves are ideal for applications in which instant on/off gas control is necessary.
  • Rated to 200 psig at 21 °C
  • Maximum temperature range of about 148 °C