Ball valve, Parker Balston® shut-off gas valve
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
22192 22193 22194 22195
REST22192EA 0 DKK
REST22192 REST22193 REST22194 REST22195
Ball valve, Parker Balston® shut-off gas valve
Ventiler Vakuumventiler
Ball valves have a floating ball to assist sealing and to reduce operating torque, and dual seats to provide leak-tight bidirectional sealing. They are rated to 1500 psig at 21 °C and have a maximum temperature of about 177 °C.