Anion exchange chromatography columns, TSK-GEL®, DEAE
Leverandør: TOSOH Bioscience
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0007163 0007164 0007168 0007210 0007574 0007648 0008802 0008806 0014016 0014466 0018757 0018761 0016092
TOYO0007163EA 13180 DKK
TOYO0007163 TOYO0007164 TOYO0007168 TOYO0007210 TOYO0007574 TOYO0007648 TOYO0008802 TOYO0008806 TOYO0014016 TOYO0014466 TOYO0018757 TOYO0018761 TOYO0016092
Anion exchange chromatography columns, TSK-GEL®, DEAE
Kromatografikolonner
TSK-Gel® DEAE is a strong anion exchange resin composed of highly cross-linked polymethacrylate beads for bimolecule purification.