1-(Cyclopropylcarbonyl)-4-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]piperazine ≥98%
Leverandør: Cayman Chemical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Olaparib, 4-(3-(4-(Cyclopropanecarbonyl)piperazine-1-carbonyl)-4-fluorobenzyl)phthalazin-1(2H)-one
10621-25 10621-5 10621-100 10621-10
CAYM10621-25EA 3000 DKK
CAYM10621-25 CAYM10621-5 CAYM10621-100 CAYM10621-10
1-(Cyclopropylcarbonyl)-4-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]piperazine ≥98%
1-(Cyclopropylcarbonyl)-4-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]piperazine
Olaparib is a potent inhibitor of PARP1 and PARP2 (IC50 = 5 and 1 nM, respectively) but is less effective against the PARP tankyrase-1 (IC50 = 1,5 µM). It can be used in cells and in animals, alone or in combination therapy with alkylating agents, to block BER and increase cancer cell death.
These products generated by alkylating agents on DNA can be efficiently repaired by normal base excision repair (BER). Some poly(ADP-ribose) polymerases (PARPs) assist in the repair of single-strand DNA nicks, an important step in BER.
Formel: C₂₄H₂₃FN₄O₃
Molekylvægt: 434.47 g/mol
MDL Number: MFCD13185161
CAS-nummer: 763113-22-0