Precleaned volatile organic analyte (VOA) sampling vials
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21796 21797 21798 21799 26122 26121
REST21796EA 1130 DKK
REST21796 REST21797 REST21798 REST21799 REST26122 REST26121
Precleaned volatile organic analyte (VOA) sampling vials
Hætteglas
Precleaned volatile organic analyte sampling vials with open-top caps.
  • Container, liner and closure cleaned, assembled and lot traceable
  • PTFE faced 3,2 mm silicone septa
  • Screw thread size 24 mm/400
Certificeringer: Cleaned to U.S. EPA Protocol B specifications.