DreamTaq™ Hot Start DNA polymerase
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EP1701 EP1702 EP1703 EP1704 EP1711 EP1712 EP1713 EP1714
EP1701EA 974 DKK
EP1701 EP1702 EP1703 EP1704 EP1711 EP1712 EP1713 EP1714
DreamTaq™ Hot Start DNA polymerase
Nukleinsyrereagenser End-point PCR Enzymes and Kits
Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq™ Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.
  • Higher yield of target amplicons from low template amounts
  • Increased reaction specificity due to reliable Hot Start technology
  • Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6 kb
  • Room temperature reaction set-up
  • Minimal optimisation of Mg²⁺ concentration and of primer annealing temperatures due to optimised reaction buffer
Thermo Scientific™ DreamTaq™ Green Buffer (10X) supports direct gel loading of PCR products. The two tracking dyes and a density reagent in the green buffer do not interfere with PCR performance and are compatible with downstream applications including DNA sequencing, ligation and restriction digestion.