Agarose, genetic technology grade, MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
04800568 04800668 04800669
ICNA04800568EA 4440 DKK
ICNA04800568 ICNA04800668 ICNA04800669
Agarose, genetic technology grade, MP Biomedicals
Elektroforese geler Agarose og Akrylamid
Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.
  • Free from DNAse and RNAse
  • Soluble in water (1 g/100 ml)
  • Melting point: 87,3 to 89,5 °C
Agarose is used to separate nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations. It tends to be more solid, but less elastic, than cross-linked acrylamide gels. It is also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigenMore Product Information