Tubing for rotarus® peristaltic pumps, Tygon® LFL
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
95223011 95223013 95223015 95223016 95223017
HIRS95223011EA 2840 DKK
HIRS95223011 HIRS95223013 HIRS95223015 HIRS95223016 HIRS95223017
Tubing for rotarus® peristaltic pumps, Tygon® LFL
Slanger Overførselsslange
Long flex life pump tube for general applications in the lab and industry and a long service life with maximum bending fatigue strength of all transparent Tygon® tubes, extremely low particle detachment, broad chemical resistance, long service life.
  • Temperature resistance from –54 to 74 °C
  • Hardness (Shore durometer A): 56