Tubing for rotarus® peristaltic pumps, Silicone Peroxide
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
95218036 95218035 95218013 95218017 95218016 95218015
HIRS95218036EA 0 DKK
HIRS95218036 HIRS95218035 HIRS95218013 HIRS95218017 HIRS95218016 HIRS95218015
Tubing for rotarus® peristaltic pumps, Silicone Peroxide
Slanger Overførselsslange
These silicon peroxide are for industrial use with organic peroxide, non-toxic, resistant to ozone and sunlight.
  • Temperature resistance from –51 to 238 °C
  • Hardness (Shore durometer A): 55