Tubing for rotarus® peristaltic pumps, PharMed® BPT
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
95213013 95213710 95213703 95213525 95213522 95213510 95213503 95213017 95213015 95213722 95213012 95213011 95213010 95213016 95213725 95213717
HIRS95213013EA 1800 DKK
HIRS95213013 HIRS95213710 HIRS95213703 HIRS95213525 HIRS95213522 HIRS95213510 HIRS95213503 HIRS95213017 HIRS95213015 HIRS95213722 HIRS95213012 HIRS95213011 HIRS95213010 HIRS95213016 HIRS95213725 HIRS95213717
Tubing for rotarus® peristaltic pumps, PharMed® BPT
Slanger Overførselsslange
These tubes are biocompatible for sensitive pharmaceuticaland biotechnological applications with good general chemical resistance, excellent resistance to acid, alkali and oxidation, impermeable to light and UV radiation.
  • Temperature resistance from -50 to 135 °C
  • Hardness (Shore durometer A): 64