Softwall strip cleaner, DualClean™
Leverandør: CONTEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2746S 2778 DCQT0001 DCMD0001 DCQT0002
CONT2746SEA 1300 DKK
CONT2746S CONT2778 CONTDCQT0001 CONTDCMD0001 CONTDCQT0002
Softwall strip cleaner, DualClean™
Mopper
The DualClean™ Softwall Strip Cleaner is ideal for cleaning strip curtains and barriers in all environments from clean workspace enclosure to ISO Class 5 isolated cleanroom work cells.
This is a unique system that enables one operator to easily and quickly clean both sides for 4 to 12" plastic strip curtains in a single pass. The system consists of a fabric cover that is snugly installed over a stainless steel frame with two curved-surface arms. Each curtain strip to be cleaned is positioned between the two arms of the frame, with the arms contacting both sides of the curtain at once. With slight tension maintained on teh curtain strip, the cleaner is moved down the length of the strip, easily and uniformly cleaning both sides of the entire length of the strip.