Replacement needle and plunger kits for micro-volume syringes
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
24830 24831 24833 24835 24838 24839 24834
REST24830EA 664 DKK
REST24830 REST24831 REST24833 REST24835 REST24838 REST24839 REST24834
Replacement needle and plunger kits for micro-volume syringes
Sprøjtenåle
These plunger-in-needle kits are used with micro-volume, positive displacement syringes. Plunger and needle must be replaced together.
Leveringsinformation: Kits are supplied with both a plunger and a needle.