Screw neck vials, N 18 and N 20
Leverandør: MACHEREY-NAGEL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
702098 702099
MANA702098EA 566 DKK
MANA702098 MANA702099
Screw neck vials, N 18 and N 20
Hætteglas
These vials are used for storage of liquid samples.
  • Made of clear glass
  • Flat bottom
  • 18-400 and 20-400 thread
  • Usable volume of 16 ml and 24 ml
The suitable screw closures have bonded septa.
- Septa are firmly connected with the cap (bonded)
- High quality silicone/PTFE septum materials