Replacement needles for autosampler syringes, SGE
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
24820 24821 24823 24824 24825 24826
REST24820EA 687 DKK
REST24820 REST24821 REST24823 REST24824 REST24825 REST24826
Replacement needles for autosampler syringes, SGE
Sprøjtenåle
Replacement needles for Agilent 7673, 7683, 7693A and 6850 autosampler syringes.