Tubing, stainless steel 316L, coiled
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
29071 29072 29091
REST29071EA 7850 DKK
REST29071 REST29072 REST29091
Tubing, stainless steel 316L, coiled
Slanger Overførselsslange
These sulfinert or silcosteel-CR treated tubing are recommended for inert applications, high temperatures, high pressures, corrosive environments and zero bleed.
  • High durability
Sulfinert is a required treatment for metal components when analyzing for parts-per-billion levels of organo-sulphur compounds. Silcosteel-CR a corrosion resistant layer that increases the lifetime of system components in acidic environments containing hydrochloric acid, nitric acid, or seawater.