Capillary columns, Rxi®-1HT columns (fused silica)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
13951 13952 13950
REST13951EA 6560 DKK
REST13951 REST13952 REST13950
Capillary columns, Rxi®-1HT columns (fused silica)
Kromatografikolonner
These fused silica columns are processed for high temperature applications such as high molecular weight hydrocarbons.
  • Nonpolar phase; dimethyl polysiloxane
  • Temperature range: −60 to 400 °C
Similar phases: DB-1ht, ZB-1HTinferno.