VWR®, Carboys, Wide Neck, Rectangular
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216-1907EA 791 DKK
216-1907 216-1898 216-1927
VWR®, Carboys, Wide Neck, Rectangular
Flasker Flasker, dunke
These carboys are useful for storing and dispensing solutions and media.
  • High chemical resistance
  • Ideal for sterile water storage
  • Graduated to identify the volume dispensed
PP carboys can be autoclaved before refilling to stop recurring bacterial growth. HDPE carboys are for use from –100 to +100 °C.
Note: Always remove the cap from the carboy before autoclaving.
Certificeringer: Resin meets US FDA 21 CFR, USP Class VI.