VWR®, Carboys, Round, Sanitary Neck, PP
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216-1897EA 1770 DKK
216-1897 216-1910 216-1920
VWR®, Carboys, Round, Sanitary Neck, PP
Flasker Flasker, dunke
Ideal for bioprocessing applications. Features a sanitary flange (76 mm) that accepts standard TC fittings.
  • Convenient shoulder handles
  • Very good chemical resistance
  • Autoclavable
Certificeringer: Resin meets US FDA 21 CFR, USP Class VI.