Standard microliter syringes (SGE) for Agilent 7673, 7683, 7693A, and 6850 autosamplers
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21210 21211 21212 21213 21214 21215 24780 24781 24782 24783 24784 24785 24786 24787 24790 24791 24794 24795 24799
REST21210EA 911 DKK
REST21210 REST21211 REST21212 REST21213 REST21214 REST21215 REST24780 REST24781 REST24782 REST24783 REST24784 REST24785 REST24786 REST24787 REST24790 REST24791 REST24794 REST24795 REST24799
Standard microliter syringes (SGE) for Agilent 7673, 7683, 7693A, and 6850 autosamplers
Sprøjter Sprøjter til kromatografi
SGE syringes are designed and tested to meet critical autosampler performance needs. Needle point styles are designed to withstand multiple, rapid injections through a septum.