Ball valve lockouts, standard
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
065692 065666 065669 065693
BRDY065692EA 288 DKK
BRDY065692 BRDY065666 BRDY065669 BRDY065693
Ball valve lockouts, standard
Sikkerhedslåse
These single-piece, easy to use ball valve lockouts are the ideal tool for securing quarter-turn ball valves in the OFF position.
  • With holes for either 9,53 mm or 7,00 mm padlock shackles
  • Small model: For use on ball valve sizes from 6,35 to 25,40 mm
  • Large model: For use on ball valve sizes from 31,75 to 76 mm
Max.shackle diameter: 9,53 mm or 7,00 mm
Min. shackle clearance: 19,00 mm