VWR®, Carboys, Narrow Mouth, with Cap and Sanitary Flange
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
216-1916EA 2770 DKK
216-1916 216-1926
VWR®, Carboys, Narrow Mouth, with Cap and Sanitary Flange
Flasker Flasker, dunke
Carboys, made of PP, with sanitary flange (41 mm) features welded-in closure for use as a dispensing port.
  • Moulded-in graduations
  • Very good chemical and temperature resistance
  • Handles for ease of transportation
Carboys provide a secure hook up for use as a supply reservoir to systems such as a fermentor or chromatography columns.
Certificeringer: Resin meets US FDA 21 CFR, USP Class VI.