Tubing, stainless steel 304, coiled welded/drawn
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
29238 29239 29247 29243 29245 29226
REST29238EA 4060 DKK
REST29238 REST29239 REST29247 REST29243 REST29245 REST29226
Tubing, stainless steel 304, coiled welded/drawn
Slanger Overførselsslange
These Sulfinert® treated tubings are recommended for chromatography applications, gas delivery systems, lower pressures and inert applications.
  • Stainless steel fabrication
  • Maximum temperature of 450 °C in an inert atmosphere
Sulfinert® is a required treatment for metal components when analysing for parts-per-billion levels of organo-sulphur compounds.