Test strips, water hardness, Dosatest
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
85442.601EA 234 DKK
85442.601 85443.601
Test strips, water hardness, Dosatest
Miljø test kit
A ready to use kit that fits into your lab coat pocket.
  • Rapid and convenient
  • Easy to handle
  • Choice of ranges for required precision
Ideal to monitor water hardness, especially suitable for boiler testing, aquaria and laundries. Semi-quantitative results can be obtained between 10 and 120 seconds.
The colour charts are adjusted and checked using certified standard solutions that are directly traceable to primary NIST standards.
1 mmol/litre =10 °f = 5.60 °d= 7,022 °e
1 mmol/litre= 100.09 mg/l CaCO₃
Emballering: Container with 100 test strips 6×95 mm