Manual microliter syringes, Hamilton 700 series
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
24536 24539 24967 20174 20175
REST24536EA 1170 DKK
REST24536 REST24539 REST24967 REST20174 REST20175
Manual microliter syringes, Hamilton 700 series
Sprøjter Sprøjter til kromatografi
Each syringe plunger and barrel assembly is manufactured as one working unit.
  • With cemented or removable needles
  • Components are not interchangeable or individually replaceable
Bestillingsoplysninger: Replacement cemented or removable needles available separately.