Phusion® blood direct PCR master mix
Leverandør: FINNZYMES REAGENTS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F-175L F-175S
F-175LEA 4280 DKK
F-175L F-175S
Phusion® blood direct PCR master mix
Nukleinsyrereagenser End-point PCR Enzymes and Kits
Phusion® Blood Direct PCR Master Mix is designed for amplification of DNA from whole blood. It eliminates the need for a separate DNA purification step prior to PCR. The modified Phusion® Hot Start II High-Fidelity DNA Polymerase is resistant to main PCR inhibitors present in blood and retains its activity at blood concentrations of up to 40% in the PCR reaction.
Phusion® Blood Direct PCR Master Mix is an ideal choice even for demanding applications, such as cloning. Phusion® Blood Direct PCR Master Mix also allows analysis of blood samples with different preservatives (e.g., heparin, EDTA, citrate), storage conditions and from various species.
Leveringsinformation: Kit contains Phusion® Blood Direct PCR Master Mix (2X), universal control primer mix, 50 mM EDTA, 50 mM MgCl2, DMSO, nuclease-free water, and O'GeneRuler Express DNA ladder.