Agarose, electrophoresis grade, MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
04820721 04820723
ICNA04820721EA 3740 DKK
ICNA04820721 ICNA04820723
Agarose, electrophoresis grade, MP Biomedicals
Elektroforese geler Agarose og Akrylamid
Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.
  • Free from DNAse and RNAse
  • Soluble in water
  • Melting point: 87,3 to 89,5 °C
Agarose is useful in separation of nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations and in chromatographic separations by either beading the agarose or cross-linking it. Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.