Microorganism tests, preservation system, CryoInstant
Leverandør: DELTALAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
409113/1 409113/2 409113/3 409113/4 409113/5 409113/6
DELT409113/1EA 2870 DKK
DELT409113/1 DELT409113/2 DELT409113/3 DELT409113/4 DELT409113/5 DELT409113/6
Microorganism tests, preservation system, CryoInstant
Mikrobiologiske testsystemer Test for mikroorganismer
CryoInstant is a preservation system for storing microbiological culture consisting on 2 ml cryovials containing 25 porous beads and cryopreservative-added broth.
  • Perfect freezing storage system (Save space in the freezers when storing)
  • Can be used to obtain up to 25 replicates from the original microorganism (to use them during many years)
  • Ease the recovery of the stored strain streaking an inoculated bead directly onto solid media
  • Prevent changes and repetitive thawing/freezing of the rest of beads
  • Avoid the growth of ice when recovering
  • Improve recovery rates of frozen cultures by adding cryopreservative to broth
  • Minimise the risk of cross contamination
The external thread cryovials are made of PP with smooth inner surface which prevents from
contamination. Their caps embody a silicone washer to ensure a positive leakproof seal. Tubes and caps withstand up to –190 ºC.
Emballering: Cryovials are packed in carton boxes with 100 units. Each box is supplied plastic-wrapped and labelled with code reference, batch, expiration date and specifying the colour of the caps and beads.