Packed GC columns, 2abc refinery column set
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
88000-875 88004-875 88000-810
REST88000-875EA 16370 DKK
REST88000-875 REST88004-875 REST88000-810
Packed GC columns, 2abc refinery column set
Kromatografikolonner
Special columns for unsaturated light hydrocarbons.
  • Faster separations of C1 to C4 hydrocarbons
When used in the proper valving system, it will elute C5+ hydrocarbons ahead of C1 through C4 hydrocarbons.