Sampling bottles, wide mouth, HDPE, with screw caps
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8126H-30 8126H-500 8126H-125 8126H-250 8126H-60
SAMP8126H-30EA 673 DKK
SAMP8126H-30 SAMP8126H-500 SAMP8126H-125 SAMP8126H-250 SAMP8126H-60
Sampling bottles, wide mouth, HDPE, with screw caps
Flasker Plastflasker
These wide mouth bottles have a screw on cap and are ideal for transporting samples, laboratory work and general use in production. The wide opening means that these bottles are suitable for both powders and liquids.
  • HDPE body with PP lid
  • Robust, water tight construction
  • Translucent HDPE walls enable users to see the material inside
  • Wide mouth means they can be filled easily
  • Made from food grade and FDA compliant material
  • BSE/TSE free
Certificeringer: Conforms to the latest FDA and EU requirements. USP class VI compliant.