Y-27632, Millipore®
Leverandør: Merck Millipore Calbiochem
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: trans-4-((R)-1-Aminoethyl)-N-(pyridin-4-yl)cyclohexanecarboxamide
688000-1 688000-5 688000-10 688000-100MG
688000-1EA 3420 DKK
688000-1 688000-5 688000-10 688000-100MG
Y-27632, Millipore®
Y-27632
A highly potent, cell-permeable, reversible, and selective inhibitor of Rho-associated protein kinases (Ki = 140 nm for p160ROCK). Also inhibits ROCK-II with equal potency. Also acts as a potent inhibitor of agonist-induced Ca2+ sensitization of myosin phosphorylation and smooth muscle contraction.
  • Inhibition is competitive with respect to ATP
  • Exhibits 10- to 50-fold lower affinity for PKCε than p160ROCK
  • Pretreatment with Y-27632 reduces the risk of lung injury in rat models of sepsis
  • Does not affect the activity of p21-activated protein kinase (PAK) even at higher concentrations (~100 µm)
Prevents apoptosis and enhances the survival and cloning efficiency of dissociated hES cells without affecting their pluripotency. A 5 mm (500 µg/296 µl) solution of Y-27632 in H2O and a 10 mm solution in DMSO are also available.
Formel: C₁₄H₂₁N₃O
Molekylvægt: 247.34 g/mol
MDL Number: MFCD03093855
CAS-nummer: 146986-50-7