Inlet liner packing, CarboFrit™
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20294 20295
REST20294EA 575 DKK
REST20294 REST20295
Inlet liner packing, CarboFrit™
Filtre Forkolonnefiltre og fritter
CarboFrit™ plugs are specifically designed to be slightly larger in OD than the 4 mm ID of glass liners. This allows the plug to conform to the ID of the glass liner, preventing voids in the packing.
For easier installation, and to reduce the possibility of flaking or creating particle debris, moisten the CarboFrit™ plug with methanol or methylene chloride before installing it into the liner.