Bulb pipettes, with individual certificate, BLAUBRAND®, two marks, Class AS
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
929734 929735 929738 929722 929723 929727 929721 929725 929732 929733
BRND929734EA 427 DKK
BRND929734 BRND929735 BRND929738 BRND929722 BRND929723 BRND929727 BRND929721 BRND929725 BRND929732 BRND929733
Bulb pipettes, with individual certificate, BLAUBRAND®, two marks, Class AS
Pipetter Kolbepipetter
AR-Glas®
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Including an individual certificate
Certificeringer: DIN EN ISO 648; DE-M marking