Nitrile gloves, uvex u-fit
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6059609 6059608
UVEX6059609EA 192 DKK
UVEX6059609 UVEX6059608
Nitrile gloves, uvex u-fit
Handsker
These disposable safety gloves provide reliable spray protection when handling chemicals such as acids, alkalis, solids or aqueous saline solutions.
  • Good grip with the roughened surface
  • Very good abrasion resistance
  • Silicone-free according to imprint test
  • Outstanding tactile feel
  • Very good fit
  • Extremely high flexibility
Typical applications include: Precision assembly work, inspection, short periods handling chemicals, paint shop (as splash protection), food processing, and gentle cleaning.
Certificeringer: EN ISO 374-1 Type B (KPT); AQL 1,5
Food contact approved