Industrial gloves, uvex contact ergo ENB20C
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6015000 6015001 6015002 6015003 6015004 6015099
UVEX6015000EA 376 DKK
UVEX6015000 UVEX6015001 UVEX6015002 UVEX6015003 UVEX6015004 UVEX6015099
Industrial gloves, uvex contact ergo ENB20C
Handsker
These cotton interlock liners feature a special NBR coating (nitrile rubber) on palm and fingers. Ideal for use in oily and greasy areas of application.
  • Thick, hard-wearing
  • Very good grip in damp, wet and oily areas
  • Good tactile feel
  • Ergonomic fit, highly flexible
  • Good wearer comfort with high water vapour absorption of the cotton lining
Typical applications include: Assembly, inspection, repair work, shipping/logistics and packaging.
EN 388 (2121)
Food contact approved
Certificeringer: EN 388:2016 (2121X)
Food contact approved
OEKO-TEX® Standard 100