Tubing, fused silica
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10050 10051 10052 10055 10056 10057 10060 10061 10062
REST10050EA 858 DKK
REST10050 REST10051 REST10052 REST10055 REST10056 REST10057 REST10060 REST10061 REST10062
Tubing, fused silica
Slanger Kromatografirør
Fused silica tubings are strength tested and economical. They are coiled in 7" diameters, banded in 5-meter sections and mounted in a cage.