Desiccator cabinet, SICCO star vitrum horizontal
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
V1821-07 V1825-07
BOHLV1821-07EA 20440 DKK
BOHLV1821-07 BOHLV1825-07
Desiccator cabinet, SICCO star vitrum horizontal
Ekssikatorer Ekssikatorkabinetter
Transparent cabinets, have an reinforced aluminium frame with panels made of heat-resistant borosilicate glass 3.3. Suitable for storing hot materials (max. 300 °C). The units have very good resistance compared to most acids and organic substances.
  • Door with magnetic catch and circular rubber seal
  • Variable height stainless steel shelves
  • Easy to read electronic hygrometer
  • Temperature resistance from -70 to +150 °C
  • Cabinets have non slip rubber feet and arestackable
Leveringsinformation: Supplied with two shelves made of aluminium, tray, hygrometer andMore Product Information