Corning® Treated Culture Dishes
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
430165 430166 430167 430196 430293 430599 431110 3261 3295 4615
734-1698EA 2200 DKK
734-1698 734-1699 734-1815 734-1703 734-1705 734-1711 734-1727 734-0884 734-4056 CORN4615
Corning® Treated Culture Dishes
Dyrkningsplader Celledyrkningsskåle
Optisk klar PS. Kan fås med behandlet overflade. Corning® CellBIND®-overfladebehandling gør overfladen mere hydrofil for at give en mere jævn og ensartet binding af celler. Ultra-Low Attachment-skåle er forsynet med et kovalent bundet hydrogel-lag, som minimerer binding af celler, proteinabsorption og cellulær aktivering.
  • Med stablingsring, som letter håndteringen
  • Leveres med ventilationshuller for at give ensartet gasudveksling
  • Steriliseret vha. gammabestråling
  • Certificeret non-pyrogen
These disposable petri-style dishes are manufactured from optically clear virgin polystyrene. Dishes are equipped with stacking beads to aid in handling and feature vents to ensure consistent gas exchange. Sterilized by gamma irradiation.
The surface minimizes cell attachment, protein absorption, and enzyme activation. It is ideal for culturing stem cells, as it promotes ebryoid body formation from ES cells. Surfaces are stable, noncytotoxic, biologically inert, and nondegradable.