BioAssay-skåle, 245×245 mm, kvadratiske, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
431111 431272 431301
734-1728EA 1910 DKK
734-1728 734-1731 734-1732
BioAssay-skåle, 245×245 mm, kvadratiske, Corning®
Dyrkningsplader Skåle til biologiske analyser
Optically clear PS. For applications where cell attachment is not desired.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilized by gamma radiation
  • Certified non-pyrogenic
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning
Miljøcertificering RECs, ISO 14001 - Oneonta