Sporicid, Actril® middel til koldsterilisering
Leverandør: MARCOR PURIFICATION
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
176-02-053 176-02-056
148-0106EA 5730 DKK
148-0106 148-0111
Sporicid, Actril® middel til koldsterilisering
Renseopløsninger Disinfektanter
Actril® Cold Sterilant is a ready to use, effective, viricide, bactericide, fungicide, and sporicide for surface disinfection in pharmaceutical environments. It is completely biodegradable and contains only pharmaceutical grade raw materials.
  • No dilution required
  • Bactericidal, mycobactericidal, fungicidal, virucidal and sporicidal
  • Faster, safer, and less corrosive than aldehyde-based liquids
  • Filtered through 0,2 µm filters
Effective ready-to-use sporicide is available in cleanroom packaging to make it easier to use in cleanroom settings. Actril® Cold Sterilant is a leading surface disinfectant in medical and cleanroom applications. Actril® Cold Sterilant complies with the USP guidelines for choosing disinfectants for aseptic areas. Actril®CR is completely biodegradable and does not have to be part of a rotation of disinfectants program.