Steriliseringsposer
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
759705 759710
129-0131EA 192 DKK
129-0131 129-9812
Steriliseringsposer
Poser Affaldsposer
Bags: Disposal bag, PP/PA.
Holders:
Poseholdere fremstillet af stålwire belagt med epoxyharpiks. Gummifod. Ikke autoklaverbar.
Epoxy-coated steel wire, with rubber feet. Non-autoclavable.
Advarsel: risk of injury. Never put sharp objects such as needles into plastic bags. Bags should stay open during sterilisation.