GC Columns, Rt-bDEXse™ (fused silica)
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
13106 13107
REST13106EA 8120 DKK
REST13106 REST13107
GC Columns, Rt-bDEXse™ (fused silica)
Kromatografikolonner
Separations of more than one hundred chiral compounds have been achieved using our unique DEX columns, and DEX columns continue to demonstrate stability after hundreds of temperature program cycles. By adding β or γ cyclodextrin to our bonded Rtx-1701 stationary phase, we greatly enhance overall utility and column lifetime for our chiral columns, compared to columns that have pure cyclodextrin stationary phases. 2,3-di-O-ethyl-6-O-tert-butyl dimethylsilyl beta cyclodextrin added into 14% cyanopropylphenyl/86% dimethyl polysiloxane