GC Columns, Rt-bDEXsm™
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
13104 13105
REST13104EA 8120 DKK
REST13104 REST13105
GC Columns, Rt-bDEXsm™
Kromatografikolonner
Separations of more than one hundred chiral compounds have been achieved using our unique DEX columns, and DEX columns continue to demonstrate stability after hundreds of temperature program cycles. 2,3-di-O-methyl-6-O-tert-butyl dimethylsilyl beta cyclodextrin added into 14% cyanopropylphenyl/86% dimethyl polysiloxane.These are excellent column for most chiral compounds in essential oils.