GC Columns, Rt-bDEXm™
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
13100 13101
REST13100EA 5730 DKK
REST13100 REST13101
GC Columns, Rt-bDEXm™
Kromatografikolonner
Separations of more than one hundred chiral compounds have been achieved using our unique DEX columns, and DEX columns continue to demonstrate stability after hundreds of temperature program cycles. Permethylated beta cyclodextrin added into 14% cyanopropylphenyl / 86% dimethyl polysiloxane.